Estriol

Supplier: U.S. Pharmacopeia
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

USPH1254508EA 310 GBP
USPH1254508
Estriol
Estriol
Formula: C₁₈H₂₄O₃
MDL Number: MFCD00003691
CAS Number: 50-27-1
EINECS: 200-022-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR