3-Ethyl-2-methylbenzothiazolium iodide, Sigma-Aldrich®

Supplier: SIGMA-ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

377007-5GEA 84.8 GBP
377007-5G
3-Ethyl-2-methylbenzothiazolium iodide, Sigma-Aldrich®
3-Ethyl-2-methylbenzothiazolium iodide
Formula: C₁₀H₁₂INS
MDL Number: MFCD00044134
CAS Number: 3119-93-5
EINECS: 221-494-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR