(±)-1-Benzyl-3-hydroxypiperidine, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SAFA653446-1GEA 55.9 GBP
SAFA653446-1G SAFA653446-5G
(±)-1-Benzyl-3-hydroxypiperidine, Sigma-Aldrich®
(±)-1-Benzyl-3-hydroxypiperidine
Formula: C₁₂H₁₇NO
MW: 191,27
MDL Number: MFCD00023734
CAS Number: 14813-01-5
EINECS: 238-881-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR