Tricyclohexylphosphine 20% in toluene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL455164-500GEA
446
GBP
SIAL455164-500G
SIAL455164-100G
Tricyclohexylphosphine 20% in toluene, Sigma-Aldrich®
Tricyclohexylphosphine
Formula:
C₁₈H₃₃P MW: 280.43 g/mol |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 EINECS: 220-069-2 UN: 1294 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...