Bis(tetramethylcyclopentadienyl)manganese(II), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Bis(tetramethylcyclopentadienyl)manganese
SIAL562424-1GEA
190
GBP
SIAL562424-1G
Bis(tetramethylcyclopentadienyl)manganese(II), Sigma-Aldrich®
Bis(tetramethylcyclopentadienyl)manganese (II)
Formula:
C₁₈H₂₆Mn MW: 297.34 g/mol |
MDL Number:
MFCD01862454 CAS Number: 101932-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...