Thallium(I) acetylacetonate, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Thallium(I) 2,4-pentanedionate

SAFA153885-5GEA 181 GBP
SAFA153885-5G
Thallium(I) acetylacetonate, Sigma-Aldrich®
Thallium (I) acetylacetonate
Formula: C₅H₇O₂Tl
MW: 303,491 g/mol
MDL Number: MFCD00013506
CAS Number: 25955-51-5
EINECS: 000-000-0
UN: 1707
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR