Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) ≥99.9% (metals basis)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Co(TMHD)3
, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)cobalt(III)
41280.03EA
75
GBP
41280.03
41280.06
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) ≥99.9% (metals basis)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III)
Formula:
C₃₃H₅₇CoO₆ MW: 608.735 g/mol Boiling Pt: 120 °C (0.5 mmHg) (subl.) Melting Pt: 254…256 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233617 CAS Number: 14877-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...