Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II), Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II)
, Ni(TMHD)2
M-Clarity™ quality level = MQ100
SIAL403393-1GEA
118
GBP
SIAL403393-1G
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II), Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II)
Formula:
C₂₂H₃₈NiO₄ MW: 425.23 g/mol |
MDL Number:
MFCD00192348 CAS Number: 14481-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...