Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')niobium(IV) ≥99%
Supplier: ABCR GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)niobium(IV)
, Nb(TMHD)4
ABCRAB121263.0005EA
0
GBP
ABCRAB121263.0005
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')niobium(IV) ≥99%
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')niobium(IV)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₄₄H₇₆NbO₈ MW: 825.976 g/mol Boiling Pt: 325 °C (dec.) Melting Pt: 219…220 °C |
MDL Number:
MFCD00145379 CAS Number: 41706-15-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...