Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV) ≥99.9%
Supplier: Alfa Aesar
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Zr(TMHD)4
, Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV)
39548.03EA
32.9
GBP
39548.03
39548.06
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV) ≥99.9%
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zirconium(IV)
Formula:
C₄₄H₇₆O₈Zr MW: 824.29 g/mol Boiling Pt: 180 °C (0.1 mmHg) (subl.) Melting Pt: 318…320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00145380 CAS Number: 18865-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...