Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II), Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)zinc(II)Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)zincZn(TMHD)2

SAFA417734-1GEA 86.3 GBP
SAFA417734-1G
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II), Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')zinc(II)
Formula: C₂₂H₃₈O₄Zn
MW: 431,92 g/mol
MDL Number: MFCD00192578
CAS Number: 14363-14-5
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR