Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III), Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: La(TMHD)3Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III)

SAFA237264-1GEA 87.2 GBP
SAFA237264-1G
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III), Sigma-Aldrich®
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')lanthanum(III)
Formula: C₃₃H₅₇LaO₆
MW: 688,71 g/mol
MDL Number: MFCD00010466
CAS Number: 14319-13-2
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR