Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)erbium(III)Er(TMHD)3

SIAL434078-1GEA 80.9 GBP
SIAL434078-1G
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III), Sigma-Aldrich®
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')erbium(III)
Formula: C₃₃H₅₇ErO₆
MW: 717.061 g/mol
MDL Number: MFCD00015705
CAS Number: 35733-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR