Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III) ≥98%

Supplier: Alfa Aesar
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)dysprosium(III)Dy(TMHD)3

36464.02EA 13.2 GBP
36464.02 36464.04
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III) ≥98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')dysprosium(III)

Superconductor MOCVD Precursor

Formula: C₃₃H₅₇DyO₆
MW: 712,302 g/mol
Melting Pt: 182…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00000017
CAS Number: 15522-69-7
EINECS: 239-563-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR