Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium, Sigma-Aldrich®

Supplier: Sigma-Aldrich
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium(II)

SAFA697524-1GEA 64.5 GBP
SAFA697524-1G
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium, Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium
Formula: C₂₂H₃₈O₄Sr
MDL Number: MFCD00064758
CAS Number: 36830-74-7
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR