Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)barium hydrate
, [Ba(TMHD)2]
SIAL362948-5GEA
426
GBP
SIAL362948-5G
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate, Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium hydrate
Formula:
C₂₂H₃₈BaO₄·H₂O MW: 521.877 g/mol |
MDL Number:
MFCD06200255 CAS Number: 17594-47-7 UN: 1564 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...