Water baths with circulation system
Supplier: GFL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
462-5121EA 1480 GBP
462-5121 462-5122
Water baths with circulation system
Baths Water Baths, Stirred
The circulation system equipped in these water baths helps to maintain an optimal temperature uniformity. Bath interior, heating element, lid, cover frame and perforated floor made of stainless steel. The outer housing of the system is corrosion resistant and made of powder-coated, electrolytically galvanised sheet steel.
  • Short heating-up times
  • Electronic monitoring of the temperature regulation
  • Soft-touch keys with clear symbols
  • Cover frame and lid have mirror finish
An electric motor with rotary magnet is flanged to the bath floor which transmitted torque to the PTFE coated stirring magnet and hence the water flow in the bath occurs.
Used for incubations and inactivation of cultures, such as for warming bacteriological media, carrying out chemical reactions or thawing samples.