Pipette controller, manual, Profiller™ 435
Supplier: Socorex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Profiller™
SOCO435.100EA 74.3 GBP
SOCO435.100
Pipette controller, manual, Profiller™ 435
Pipettes Pipette Controllers
This manual, easy to use pipette controller provides smooth control in both aspirating and dispensing, using glass or plastic pipettes of volume up to 100 ml.
  • Large capacity bulb
  • Soft action lever button and efficient blow out system
  • PTFE protection membrane filter
  • Maintenance free
  • Alternative colour coded nose cones available