Sutures, Premilene®
Supplier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Premilene®
BRAUC2090619EA 828 GBP
BRAUC2090619
Sutures, Premilene®
Sutures
Premilene® is composed of an isotactic polypropylene, which presents an absolutely regular and longitudinal molecular structure.
  • Prolonged tensile strength retention
  • Adequate elasticity and elongation values, which guarantee a stable and secure knot
  • Pliability for easy knot tying and comfortable continuous suturing
  • Strong, smooth, and flexible monofilament
  • Circle reverse cutting needle and micro-point DSMP 13