Would you like to log in again?
Round tissue float bath
Supplier: GFL (GESELLSCH. FÜR LABORT.)
631-1300
Round tissue float bath
The tissue float bath has a black anodised aluminium interior, the housing is powder coated aluminium. The unit has precise temperature regulation which ensures evenly stretched paraffin wax sections, and allows the sections to be easily seen in the bath or on the rim of the bath when drying.
  • Low height (100 mm) enables comfortable and safe working
  • Knob and scale temperature regulator
  • Control thermometer in stainless steel holder is fixed to the rim of the bath
Ordering information: Supplied without aluminium lid which protects against dust.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty