Half masks against gas, vapour and dust, 8000 series
Supplier: Moldex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
111-1177EA 32.6 GBP
111-1177 111-1178 111-1179
Half masks against gas, vapour and dust, 8000 series
Respirators Reusable Respirators Half Face Respirators
These versatile respirators can be used for gas/vapour protection, gas/vapour plus particulate protection, or only particulate protection dependent upon which combination of adapters and cartridges are used. The mask body is made from soft, durable TPE material.
  • Easy maintenance and cleaning without tools
  • Radial connector: New inhalation valve with every cartridge
  • Improved field of vision, lightweight and easy to fit
Certifications: EN 140:1998