Flow meter tubes for air
Supplier: GLASS PRECISION ENGINEERING
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GLPR10414EA 137 GBP
GLPR10414 GLPR10417 GLPR10423 GLPR10328 GLPR10315 GLPR10405 GLPR10408 GLPR10420 GLPR10311 GLPR10323 GLPR10303 GLPR10309 GLPR10310 GLPR10322 GLPR10411 GLPR10314 GLPR10302 GLPR10425 GLPR10407 GLPR10401 GLPR10307 GLPR10426 GLPR10424 GLPR10416 GLPR10325 GLPR10304 GLPR10427 GLPR10403 GLPR10321 GLPR10306 GLPR10320 GLPR10413 GLPR10419 GLPR10305 GLPR10317 GLPR10324 GLPR10422 GLPR10412 GLPR10312 GLPR10421 GLPR10319 GLPR10318 GLPR10406 GLPR10316 GLPR10404 GLPR10428 GLPR10409 GLPR10308 GLPR10415 GLPR10410 GLPR10313 GLPR10418 GLPR10326 GLPR10402 GLPR10327 GLPR10301
Flow meter tubes for air
Flow Meters
Borosilicate glass tubes with linear or direct reading scale giving air or water flow measurements. Clear indelibly fired-in markings and yellow backing give ease of reading. The tubes are made of heat-resisting glass with heavy walls.
  • Repeatability ±0,25 % of full scale reading
  • Rib guided flow tube for maximum stability and accuracy
  • Polyethylene connector
Delivery information: Supplied with push-on plastic tube connector.