Would you like to log in again?
PCR plates, 96-well, semi-skirted, raised skirt, barcoded
Supplier: Thermo Scientific
ABgene®
PP. These 96-well semi-skirted plates offer increased rigidity, which allows for multi-channel pipetting and reduces plate warping during thermal cycling applications. These plates feature a cut corner at the A12 location and thin well walls. They are compatible with all ABI platforms, including sequencers, and do not require adapters.
  • Directly compatible with all ABI platforms, including sequencers, with no adapters necessary
  • Available as SuperPlate providing 4× more rigidity for superior robotic handling
  • Cut corner (A12)
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer
  • White plates are optimised for qPCR by increasing sensitivity and reducing variability in fluorescence detection
The barcodes are the industry standard code 128. The label itself can withstand temperatures from −196 ºC up to 120 ºC to ensure that the barcodes stay on the plate throughout the entire workflow. It also has a special coating to virtually eliminate any ‘rubbing off’ of the code during handling. The dual barcodes are located on the opposite numeric and opposite alpha sides of the plate.
ORDER
Expand All / Collapse All
Description Frame colour Well colour Pk VWR Catalog Number Quantity
96-well PCR SuperPlate, semi-skirted with raised deck, barcoded White White 25 732-1493
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty