MaxiMix™ III vortex mixer combines a heavy, cast-aluminum base with counterbalance system for strength and stability.
- Precise matching of an electronic speed control to a high torque DC type motor provides versatile speed selection ranging from 100 to 2200 rpm
- Soft foam rubber top (15,8×13,9 cm) allows vortexing multiple test tubes and small flasks
- Lighted main power switch indicates mixer is operating
- High-torque DC motor for consistent mixing action
- Spring-dampened feet eliminate 'creeping and walking' during large volume mixing
- Continuous trouble-free shaking of loads up to 2,2 kg
MaxiMix™ III can perform the functions of four separate apparatuses and save valuable bench space.
Ordering information: The unit is supplied complete with one soft foam top.