Temperature logger, Junior
Supplier: Escort
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Junior
ESCOEJ-EG-B-16EA 152 GBP
ESCOEJ-EG-B-16
Temperature logger, Junior
Data Loggers
These loggers are designed to measure egg temperatures in cartons during transportation and storage or during the incubation process.
  • Made from ABS plastic
  • Can be placed in a standard size egg carton
  • Operate in temperatures between –20 to +60 °C
The logger has one internal sensor, which is situated in the center of the logger to simulate the response time of an actual egg. The egg logger can also be used in any other application that prohibits the use of other loggers due to size requirements.
Certifications: IP 51 rated.