Would you like to log in again?
Cleanroom wipes, Polx® 1200
Supplier: Berkshire
These knife cut unlaundered cleanroom wipes are composed of 100% continuous filament polyester knit fabric. They are designed to bridge the gap between more expensive laundered wipes and lower cost less critical non wovens
  • No binders or surfactants
  • Low contamination compared to non woven wipes
  • High abrasion resistance, good sorbency
  • Chemically compatible with IPA, Acetone and other solvents
  • Soft hand and edge for sensitive surfaces
Ideal for use in integrated circuit fabrication areas and CD/DVD manufacturing operations. Also suitable for equipment wrap, wipe downs and for applying and removing cleaning and disinfecting solutions in less critical areas.
Suitable for use in Class 100, ISO 5, M3.5 and above cleanrooms.
Colour: White
Caution: Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.
ORDER
Expand All / Collapse All
Pk VWR Catalog Number Quantity
150 BERKP1200.0909.8 Unavailable
unavailable This product is discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalogue Number listed above. If you need further assistance, please call VWR Customer Service on 0800 22 33 44.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
WRITE A REVIEW
Need help?
  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.
VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.
Please confirm the industry you work in
Example: Pharmaceutical
Please confirm your role
Example: Clinical Trial Assistant
Please confirm your location
Example: New York, NY
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty