Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Microcentrifuge tubes, 0,25/0,4 ml, with snap cap

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

MLBP3485EA 24.7 GBP InStock
MLBP3485 MLBP3483
Microcentrifuge tubes, 0,25/0,4 ml, with snap cap
Tubes Microcentrifuge Tubes
Made of LDPE (0,25 ml) or PP (0,4 ml).

  • Translucent polyethylene provides an excellent view of the sample contents
  • The smooth interior and nonwettable surface aid in complete sample removal and recovery

The specialty 0,25 ml and 0,4 ml microcentrifuge tubes are molded with an attached cap. The 0,4 ml tube is specially formulated from low-density polyethylene and can be easily cut for pellet removal.

Certifications: Non sterile: Certified DNA, Rnase/Dnase, pyrogen and endotoxin-free.
Sterile: Certified DNA, Rnase/Dnase, pyrogen, endotoxin, PCR inhibitor, and bioburden-free.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty