Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Membrane filters, Fluoropore™

Supplier: MILLIPORE
These white Fluoropore™ membrane are hydrophobic, PTFE polymer membrane bonded to a high density PE support and provide broad chemical compatibility, high flow rates and consistency.

  • PTFE with and without backing material
  • Biologically and chemically inert
  • High porosity yields high flow rates
  • Can be sterilised by ethylene oxide or autoclave
  • Available for filtration of aqueous and organic based samples

They are used for clarifying acids, bases and solvents, air monitoring, filtering and venting gases and uv spectroscopy.

Filter surface: plain

Gravimetric extractables: 0,5%

Bacterial endotoxins: 0,5 EU/ml




 

ORDER

Expand All / Collapse All
Type Pore size Ø Pk VWR Catalog Number Quantity
FGLP, white, plain 0,22 µm 13 mm 100 MILFFGLP01300
FGLP, white, plain 0,22 µm 25 mm 100 MILFFGLP02500
FGLP, white, plain 0,22 µm 47 mm 100 512-0246
FGLP, white, plain 0,22 µm 90 mm 100 MILFFGLP09050
FGLP, white, plain 0,22 µm 142 mm 50 MILFFGLP14250
FHLP, white, plain 0,45 µm 13 mm 100 MILFFHLP01300
FHLP, white, plain 0,45 µm 25 mm 100 MILFFHLP02500
FHLP, white, plain 0,45 µm 37 mm 100 MILFFHLP03700
FHLP, white, plain 0,45 µm 47 mm 100 512-0248
FHLP, white, plain 0,45 µm 90 mm 50 MILFFHLP09050
FHLP, white, plain 0,45 µm 142 mm 50 MILFFHLP14250
FALP, white, plain 1,0 µm 13 mm 100 MILFFALP01300
FALP, white, plain 1,0 µm 25 mm 100 MILFFALP02500
FALP, white, plain 1,0 µm 47 mm 100 MILFFALP04700
FALP, white, plain 1,0 µm 90 mm 50 MILFFALP09050
FALP, white, plain 1,0 µm 142 mm 50 MILFFALP14250
FALP, white, plain, with pads 1,0 µm 37 mm 100 MILFFALP03700
FSLW, white, plain 3,0 µm 25 mm 100 MILFFSLW02500
FSLW, white, plain 3,0 µm 47 mm 100 MILFFSLW04700
FSLW, white, plain 3,0 µm 90 mm 25 MILFFSLW09025
FSLW, white, plain 3,0 µm 142 mm 10 MILFFSLW14200
FHUP, unlaminated 0,45 µm 47 mm 100 MILFFHUP04700
FSLW, white, plain, with pads 3,0 µm 37 mm 100 MILFFSLW03700
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Type FALP FGLP FHLP FSLW FGLP FHLP FSLW FGLP FHLP FSLW FHLP FGLP FHUP
Pore size 1 µm 0,22 µm 0,45 µm 3 µm 0,22 µm 0,45 µm 3 µm 0,22 µm 0,45 µm 3 µm 0,45 µm 0,22 µm 0,45 µm
Thickness 150 µm 50 µm
Water flow rate 90 ml/min/cm² 15 ml/min/cm² 40 ml/min/cm² 122 ml/min/cm² 15 ml/min/cm² 40 ml/min/cm² 122 ml/min/cm² 15 ml/min/cm² 40 ml/min/cm² 122 ml/min/cm² 40 ml/min/cm² 15 ml/min/cm² 40 ml/min/cm²
Airflow rate 16 l/min/cm² 3 l/min/cm² 8 l/min/cm² 6 l/min/cm² 3 l/min/cm² 8 l/min/cm² 6 l/min/cm² 3 l/min/cm² 8 l/min/cm² 6 l/min/cm² 8 l/min/cm² 3 l/min/cm² 8 l/min/cm²
Max. temperature (°C) 130

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty