Cavity slide
Supplier: Karl Hecht
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HECH42432010EA 19.9 GBP
HECH42432010
Cavity slide
Slides and Cover Glasses Cavity Slides
This microscopy slide is made of opal glass.
  • Opal 'milk' glass
  • Frosted label
  • Marking
Suitable for blood typing.