Would you like to log in again?
VWR® VisiScope® 384 series upright trinocular microscopes
Great Deals!
VWR® VisiScope® 384 series upright trinocular microscopes
VALID UNTIL: 31/12/2017
Designed for advanced biology teaching
Complete with coated eyepieces, anti-fungus objectives plus, focusing mechanism with limit stop.
  • Quintuple nosepiece (quadruple for TL384 POL)
  • Powered LED with adjustable brightness (brightfield and phase)
  • Epi-fluorescence: High power white LED
  • Abbe condenser with iris diaphragm and graduated N.A. scale
  • Coaxial coarse and fine focusing mechanism (graduated, 0,002 mm) with upper stop
Model Contrast technique Objectives Condenser Stage Cat. No. Promo price Quantity
TL384 POL Polarisation E-plan IOS POL (strain-free) 4x/0,10; 10x/0,25; 40x/0,65; 60x/0,80 N.A. 1.25, with iris diaphragm, focusable and centrable. With rotating polarizer filter 160 mm dia.; 360° rotating with stop knob & 0,1° Vernier 630-1936E
T384FL Fluorescence & brightfield IOS E-plan achromatic 4×/0,10; 10×/0,25; 20×/0,40; 40×/0,65 & plan achromatic 50×/0,75 (no cover slide) Abbe N.A. 1,25 with centring system 188x150 mm (X/Y movement: 78x54 mm) 630-1937E
TL384PHI Phase contrast, darkfield (dry) & brightfield IOS plan achromatic 10xPh, 20xPh, 40xPh & 100xPh Phase condenser (10x/20x, 40x, 100x), with darkfield (dry) & brightfield 188x150 mm (X/Y movement: 78x54 mm) 630-2530E