Would you like to log in again?
VWR® VisiScope® IT405-FLD inverted fluo microscope
VWR® VisiScope® IT405-FLD inverted fluo microscope
VALID UNTIL: 30/09/2017
5 year warranty
This innovative tissue culture microscope, suitable for research applications. LED illumination for phase contrast, brightfield and epi-fluorescence techniques means long life and convenience.
  • Eliminate warm-up/cool-down times (60 minute time saving)
  • Pre-centred LED illumination: No frustration when changing conventional, costly halogen and mercury bulbs
  • LED lifetime over 50 000 hours
  • The lowest operational costs
  • Dual filter block for green (GFP/FITC) and red (Rhodamine/TRITC) dyes
Model Description Cat. No. Promo price Quantity
IT405-FL VisiScope® IT405-FL inverted fluo microscope 630-2695E
Epi-fluorescence camera
Pro Cool 5 Optika fluorescence camera 630-2683E
CCD798.2 Focusable C-mount adapter for 2/3” CCD sensor 630-2711E