Would you like to log in again?
VWR® VisiScope® 350 series stereo microscopes
Great Deals!
VWR® VisiScope® 350 series stereo microscopes
VALID UNTIL: 31/12/2017
Designed for advanced teaching, laboratories and industrial applications
  • WF 10×/20 - zoom 0,7× - 4,5× (for SZT350 - 6: EWF 10x/22 - zoom 0,67x - 4,5x )
  • Greenough optical system
Model Type Focusing system Working distance Illumination Cat. No. Promo price Quantity
SZB350 Binocular Pillar stand 100 mm Incident & transmitted 12 V/15 W halogen 630-1578E
SZB350 OH Binocular Stand long travel height (overhanging) 100 mm None 630-1583E
SZT350 Trinocular Pillar stand 100 mm Incident & transmitted 12 V/15 W halogen 630-1580E