Would you like to log in again?
VWR® VisiScope® 250 series upright microscopes
Great Deals!
VWR® VisiScope® 250 series upright microscopes
VALID UNTIL: 31/12/2017
Designed for both education and laboratories applications
Complete with coated eyepieces, anti-fungus objectives plus focusing mechanism with limit stop.
  • Binocular (BL) - Trinocular (TL)
  • Quadruple nosepiece
  • DIN E-plan achromatic (IOS plan achromatic for PI-model)
  • Powered LED illumination
  • Focusing coaxial coarse and fine focusing mechanism (graduated, 0,002 mm) with upper stop, to prevent contact between objective and specimen
  • Adjustable tension of coarse focusing knob
Model Objectives Condenser Stage Cat. No. Promo price Quantity
BL254 4x (N.A, 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) Sliding-in, N.A. 1,25 Abbe type with adjustable height & iris diaphragm Double layer, 150x139 mm with X-Y 75x33 mm moving range 630-1928E
BL254-PI 4x (N.A, 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) Sliding-in, N.A. 1,25 Abbe type with adjustable height & iris diaphragm Double layer, 150x139 mm with X-Y 75x33 mm moving range 630-2684E
TL254 4x (N.A, 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) Sliding-in, N.A. 1,25 Abbe type with adjustable height & iris diaphragm Double layer, 150x139 mm with X-Y 75x33 mm moving range 630-1929E