Would you like to log in again?
VWR® VisiScope® 250 series stereo microscopes
Great Deals!
VWR® VisiScope® 250 series stereo microscopes
VALID UNTIL: 31/12/2017
Designed for education
  • WF 10×/20
  • LED incident and transmitted illumination
  • Stage: Semi-transparent disc for transmitted light with specimen clips
  • Adjustable angle of incident illuminator
Model Type Objective focusing system Working distance Cat. No. Promo price Quantity
STB250 Binocular 2x, 4x Coaxial coarse & fine knobs 80 mm 630-1576E
SZB250 Binocular Zoom 0,7x - 4,5x Coarse with tension adjustment 85 mm 630-1577E
SZT250 Trinocular Zoom 0,7x - 4,5x Coarse with tension adjustment 85 mm 630-2725E