Would you like to log in again?
VWR® VisiScope® 200 series upright microscopes
Great Deals!
VWR® VisiScope® 200 series upright microscopes
VALID UNTIL: 31/12/2017
A modern and ergonomic design with reliability and innovation - perfect for teaching
Complete with coated eyepieces, anti-fungus objectives plus focusing mechanism with limit stop.
  • Monocular (ML) - Binocular (BL) - Trinocular (TL)
  • Quadruple nosepiece
  • Achromatic DIN objectives
  • LED illumination
  • Coaxial coarse and fine focusing mechanism (graduated 0,002 mm) with upper stop
Model - type Objectives Condenser Stage Cat. No. Promo price Quantity
ML214 - monocular 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 60x (N.A. 0,85) N.A. 1,25 Abbe with adjustable height & iris diaphragm Mechanical sliding stage 630-1558E
ML224 - monocular 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A 1,25) N.A. 1,25 Abbe with adjustable height & iris diaphragm Mechanical sliding stage 630-1557E
BL214 - binocular 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 60x (N.A. 0,85) N.A. 1,25 Abbe with adjustable height & iris diaphragm Mechanical sliding stage 630-1560E
BL224 - binocular 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) N.A. 1,25 Abbe with adjustable height & iris diaphragm Mechanical sliding stage 630-1559E
TL224 - trinocular 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) N.A. 1,25 Abbe with adjustable height & iris diaphragm Mechanical sliding stage 630-1859E