Would you like to log in again?
VWR® VisiScope® 100 series upright microscopes
Great Deals!
VWR® VisiScope® 100 series upright microscopes
VALID UNTIL: 31/12/2017
Basic microscope for routine biology
Complete with coated eyepieces, anti-fungus objectives plus focusing mechanism with limit stop.
  • Monocular (ML) - Binocular (BL) - with integrated camera (DML - DBL)
  • Quadruple nosepiece
  • Achromatic DIN objectives
  • LED illumination
  • Coaxial coarse and fine focusing mechanism (graduated 0,002 mm) with upper stop
Model Objectives Condenser Stage Cat. No. Promo price Quantity
BL103-POL for polarisation 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65) N.A. 1,20 with diaphragm Rotatable stage, diameter 120 mm 630-1802E
BL114 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 60x (N.A. 0,85) N.A. 1,20 with diaphragm Mechanical sliding stage 630-1553E
BL124 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) N.A. 1,20 with diaphragm Mechanical sliding stage 630-1554E
BL124-R (with rechargeable battery) 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) N.A. 1,20 with diaphragm Mechanical sliding stage 630-1926E
BL124 LED (Automatic Light Control) 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) N.A. 1,20 with diaphragm Mechanical sliding stage 630-1927E
DBL124 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A 1,25) N.A. 1,20 with diaphragm Mechanical sliding stage 630-1623E
DML103 (digital with built-in camera) 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65) N.A. 1,20 pre-centered Mechanical sliding stage 630-1556E
ML103 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65) N.A. 0,65 pre-centred Plain stage 125x120 mm 630-1550E
ML103-POL 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65) N.A. 1,20 pre-centered Rotatable stage, diameter 120 mm 630-1555E
ML114 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 60x (N.A. 0,85) N.A. 1,20 pre-centered Mechanical sliding stage 630-1551E
ML124 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) N.A. 1,20 pre-centered Mechanical sliding stage 630-1552E
ML124 LED (Automatic Light Control) 4x (N.A. 0,10), 10x (N.A. 0,25), 40x (N.A. 0,65), 100x (N.A. 1,25) N.A. 1,20 pre-centered Mechanical sliding stage 630-1925E