Would you like to log in again?
NEW VWR® reusable silicone lab mat
NEW VWR® reusable silicone lab mat
VALID UNTIL: 31/10/2017
An economical solution to help keep bench tops clean and safe from stains, spills and wear.
Save money by eliminating the need for repeated purchases and disposal costs associated with disposable paper mats. Made from a durable FDA-approved silicone material that creates a stain resistant washable working surface. When the mat is done being used, it can be easily cleaned using any standard disinfectant. Simply wipe it or putting it in a dishwasher and it’s ready for repeated use.
  • Made from FDA food grade silicone
  • Ideal for every type of laboratory and more specialized areas such as cold rooms, clean rooms, sterile suites and areas where high sterility is a requirement
  • Protect bench tops from hot items up to 200 °C (392 °F)
  • Mat can be rolled up for compact storage
  • Washable with standard disinfectants in sinks or dishwashers, allows easy cleaning up for repetitive use
Since it is reusable, it helps reduce waste making it a more environmentally friendly option over alternative paper mats. The lab mat offers a large anti-skid working surface, which when dealing with harsh chemicals and delicate glassware, helps keep items in place and stops the lab mat from sliding out from underneath.
The mat’s surface is also noise dampening, reducing sound from the vibration of common bench top equipment. It can be used with items such as centrifuges, vortex mixers, hot plates, stirrers, pipettes and so much more. When using the mat with magnetic stirrers there is a designated area with internal metal inserts to keep stir bars from wandering around the work surface. A lip-design around the outer edge of the mat helps contain spillage that may occur when in use keeping bench tops safe and clean.
Description Dimensions Depth Pk Cat. No. Promo price Quantity
Reusable silicone lab mat 300x600 mm 2 mm 1 111-9200E