Would you like to log in again?
VWR® OmniTop Sample Tubes®
VWR® OmniTop Sample Tubes®
An innovative solution from our VWR bioprocessing division
Any biopharmaceutical production requires frequent in-process sampling to confirm product integrity and conformance. To achieve this, an accurate sample must be taken that is a complete representation of the product being manufactured. But how can our customers do this quickly, cost effectively and without the risk of introducing contamination?
  • OmniTop Sample Tubes® are a convenient device that enable small volume transfers and sampling within a closed system
  • For use with bench top, pilot plant, and production bioreactors
  • Pre-sterilised, single-use
  • Available individually packaged or configured in manifold
  • See the video below for a short demonstration