Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
VWR® PCR Workstation
Contamination-free working space!
VWR® PCR Workstation
Dual surface and air cleaning action for a contamination-free working space for sensitive PCR-based applications
  • 8 mm thick, ethanol resistant safety panels absorb UV irradiation
  • Electromagnetic safety mechanism stops UV irradiation if front panel is opened
  • Work surface and rear panel made of easy to clean stainless steel
  • Function indicator for UV Air Recirculator tube
  • Displays operating time of UV tubes to enable well timed replacement of tubes for constant UV intensity
  • Out of hours programmable decontamination operation
The VWR PCR Workstation decontaminates the workspace by UV inactivation of airborne and surface-bound contaminants for preparing PCR and other samples by reducing the change of sample contamination. The unit provides effective twin action by active decontamination of the work surface during non working time by UV irradiation, plus additional inactivation of aerosol-bound contaminants by a shielded UV Air Recirculator during use.