Would you like to log in again?
VWR® 96-well low temperature PCR© rack - NEW
VWR® 96-well low temperature PCR© rack - NEW
VALID UNTIL: 31/10/2017
Unique patent-pending lid design!
The PCR© storage rack offers a sleek and modern design that’s packed full of features.
  • Patented lid: Ambidextrous, it slides out from the rack and then rotates 180° to become a convenient stand
  • Faster sample cooling: Due to the lid's side opening
  • Secure hold and sample storage: Ergonomic textured finger grips and robust ABS rack
  • Capacity: Holds 0,2 ml tubes, individual or strip, 96 well (8x12 configuration), stackable
  • For freezer storage down to -80 °C
Colour Pk Cat. No. Promo price Quantity
Assorted 5 732-2975E
Blue 5 732-3319E
Green 5 732-3320E
Purple 5 732-3321E