Would you like to log in again?
A new type of micro-sampling vial from 1,4 ml down to 20 µl (only 4 µl residual volume)
A new type of micro-sampling vial from 1,4 ml down to 20 µl (only 4 µl residual volume)
VALID UNTIL: 31/01/2018
The cost efficient alternative to a total recovery vial
  • Alternative to fused-insert vials with a bigger total max. volume but identical small residual volume
  • Self-standing conical micro-sampling vial
  • Designed for use with nearly every autosampler on the market
  • Wide range of common sample volumes can be used without changing the vial
  • Saves time and money while optimising your chromatography workflow
  Description Pk Cat. No. Promo price Quantity
  1,1 ml short thread vial, conical, glass foot, amber 1000 548-2359E  
  1,1 ml snap-ring vial, conical, glass foot, clear 1000 548-2360E  
  1,1 ml snap-ring vial, conical, glass foot, amber 1000 548-2361E  
  1,1 ml crimp-neck vial, conical, glass foot, clear 1000 548-2362E  
  9 mm caps
  Screw, PP, blue hole, red rubber 1000 548-0896E  
  Screw, PP, blue hole, silica/PTFE 1000 548-0085E  
  Screw PP, blue hole, silica/PTFE 1000 548-0088E  
  11 mm caps
  PE, blue hole, red rubber/PTFE 1000 548-3360E  
  PE snap-ring, blue hole, SI/PTFE 1000 548-0432E  
  PE snap-ring, blue hole, SI/PTFE 60° slit 1000 548-0435E  
  Combination-seal, aluminium cap with centre hole 1000 548-3272E  
  Crimp-cap, white silicone/PTFE 1000 548-0009E  
  Aluminium cap ,SI/PTFE, 60° slit, 1,5 mm cross-slit 1000 548-0411E