Would you like to log in again?
VWR® MEDical bags - NEW
VWR® MEDical bags - NEW
VALID UNTIL: 31/10/2017
The ideal solution for biological and medical sample transportation
The VWR Medbag is a liquid-tight LDPE bag, used as internal hospital and external secondary packaging for the transportation of biological substances of category B (UN 3373).
  • 100% protection: Unique opening system gives perfect leak resistance which cannot be achieved with a pre-cut opening perforation
  • Unique identification: Track & Trace technology avoids sample confusion
  • Easy to close/open: Guarantees a perfect seal with a fast adhesive closure and a quick and easy opening
  • Compliance: UN 3373; ADR 2013/P650 WHO/HSE/GCR/2012.12 - ISO 15189
  • Biohazard logo and instructions for use printed on bag
Description Dimensions Pouch type Application Pk Cat. No. Promo price Quantity
Medbag biological sampling bag, small 176x230 mm Patient information record sheet has to be folded in three and inserted into the outer pouch. External laboratories 2000 129-1175E
Medbag biological sampling bag, medium 176x270 mm Patient information record sheet can be folded in half and inserted into the outer pouch which has an additional flap to ensure the document is held secure. Hospitals 2000 129-1176E
Description Capacity Application Pk Cat. No. Promo price Quantity
Absorbent pads 25 ml Designed to ensure safe and proper sample storage and transport. They absorb the spills in case of accidental damage. 2000 129-1197E