Would you like to log in again?
LABWASH® Premium detergent range
LABWASH® Premium detergent range
VALID UNTIL: 31/10/2017
Efficient, ecological detergent range for cleaning your laboratory utensils without residue
A new range of 20 different detergents to clean your laboratory from top to bottom.....
  • High quality
  • Excellent power of cleaning with no residue
  • High activity and low consumption
  • High compatibility with most materials
  • Ecological with high purity active biodegradable ingredients
Description Specifications Pk Cat. No. Promo price Quantity
LABWASH® Premium Alkamatic Mildly alkaline, liquid cleaning concentrate for special washing machines 5 l 84549.360E
LABWASH® Premium Alkamatic PF Alkaline, liquid cleaning concentrate for special washing machines (ideal for heavy contamination, phosphate- and chlorine-free) 5 l 84550.360E
LABWASH® Premium Alkamatic LA Highly alkaline intensive cleaner for laboratory washers 5 l 84556.360E
LABWASH® Premium Acidrinse C Phosphate-free neutraliser for special washing machines 5 l 84551.360E
LABWASH® Premium Acidrinse P Neutraliser and pre-cleaner for special washing machines 5 l 84552.360E
LABWASH® Premium Easyrinse Acidic, ecological rinsing concentrate with shiny effect 5 l 84553.360E
LABWASH® Premium Pure Concentrate for soak and spray cleaning of laboratory glassware 5 l 84555.360E