Would you like to log in again?
VWR® INCU-Line® Prime incubators
Great Prices!
VWR® INCU-Line® Prime incubators
VALID UNTIL: 31/12/2017
Optimal environment for your samples with superior temperature uniformity
Natural convection incubators with stainless steel chamber inside and internal glass door.
  • Microprocessor controller with LCD graphic display, stores up to 3 user programs
  • USB interface allows download of relevant program data with Basic Control Software
  • Real time clock, door lock and access port (Ø = 30 mm)
  • Manual air flap control
  • Over-temperature protection (safety thermostat class 3.1)
Supplied with 2 stainless steel wire shelves for 56 and 112 Prime models and 3 shelves for the 180 Prime model. Manufacturer test certificate (data measured at 37 °C) and Basic Control Software to download data from internal memory.
Model IL 56 IL 112 IL 180
Temp. range (°C)       Ambient +5...+100
Capacity (l) 56 112 180
Temp. variation (spatial) (°C) at 37 °C ±0,7 ±0,6 ±0,8
Temp. fluctuation (time) (°C) at 37 °C                           ±0,2    
Shelves supplied (max.) 2 (5) 2 (7) 3 (9)
Max. load per shelf/max. per unit (kg) 25/50 25/50 25/75
W×D×H int. (mm) 395×360×395 460×450×540 465×560×720
W×D×H ext. (mm) 595×600×700 655×700×845 650×760×1030
Weight (kg) 52 69 94
Power consumption (W) 450 450 850
Description Cat. No. Promo price Quantity
INCU-Line® IL 56 Prime 390-0909E
INCU-Line® IL 112 Prime 390-0910E
INCU-Line® IL 180 Prime 390-0911E
Description Cat. No. Price Quantity
Stainless steel shelf for IL 56 466-0454E
Stainless steel shelf for IL 112 466-0455E
Stainles steel shelf for IL 180 466-0456E
Base on castors for IL 180 (H: 47 mm) 466-0457E