Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.
VWR® INCU-Line IL 68R cooled incubator
Incubator -20%!
VWR® INCU-Line IL 68R cooled incubator
VALID UNTIL: 31/12/2017
Cooled incubator with forced convection
Ideal for processes and tests at, or below, ambient temperatures.
  • Adjustable hold at set point time for temperature from 1 minute to 31 days or continous operating
  • Microprocessor controller with LCD graphic display can store up to 3 user programs
  • USB interface to download data from internal memory with Basic Control Software
  • External glass door, 30 mm access port, internal LED light with real time clock and door lock
  • Stainless steel interior, internal socket (EU plug only) and power failure control system
  • Over-/under-temperature protection (safety thermostat class 3.3)
Unit is suitable for BOD measurement but not stackable. Supplied with two coated wire shelves, temperature test certificate (data measured at 37 °C) and Basic control software on CD.
Cap. (l) 68
Temp. range (°C) +3…70
Temp. resolution (°C) 0,1°C
Temp. variation (spatial) (°C) ±0,3 at 37 °C
Temp. fluctuation (time) (°C) ±0,1 at 37 °C
Ext. WxDxH (mm) 570x670x600
Int. WxDxH (mm) 470x300x430
Power consumption (W) 160
Weight (kg) 38
Description Cat. No. Promo price Quantity
INCU-Line® IL 68R cooled incubator 390-0728E
Description Cat. No. Price Quantity
Additional shelf for IL 68R 390-0733E