Would you like to log in again?
VWR® INCU-Line® CR Premium cooled incubators
20% off the incubators
VWR® INCU-Line® CR Premium cooled incubators
VALID UNTIL: 31/12/2017
Large incubators with compressor cooling and forced convection
Complete with an acid-proof stainless steel (DIN 1.4301) inner chamber and a powder-coated sheet exterior. The adjustable heating and cooling system provides a temperature range between –10 to +100 °C. All models have a solid outer door and an internal glass door. The larger IL 400CR and IL 750CR models have double outer doors. All models are equipped with levelling castors, 30 mm access port and internal light.
  • Full colour LCD touch screen, displays values in tabular or graphic form
  • Access control via login, 20 user-program memory and 7-day program mode
  • USB port to save data on flash drives, LAN interface, remote control via Internet and Ethernet cable
  • Over-/under-temperature protection (safety thermostat class 3.3)
All models are supplied with 3 stainless steel wire shelves, test certificate (data measured at 37 °C), Ethernet cable and PREMIUM Control Software. 
Untitled Document
Model IL 240CR Premium IL 400CR Premium IL 750CR Premium
Temp. range (°C)       -10 to +100
Capacity (l) 245 424 749
Shelves supplied (max.) 3 (10) 3 (14) 3 (16)
Max. load per shelf/max. per unit (kg) 25/90 25/120 25/140
W×D×H int. (mm) 600×510×800 800×510×1040 1040×600×1200
W×D×H ext. (mm) 820×760×1430 1040×740×1650 1260×860×1820
Weight (kg) 140 185 275
Power consumption (W) 800 1200 1800
Temp. variation (spatial) (°C) at 37 °C ±0,3   
Temp. fluctuation (time) (°C) at 37 °C ±0,2    
Description Cat. No. Promo price Quantity
INCU-Line® IL 240CR Premium 390-0964E
INCU-Line® IL 400CR Premium 390-0965E
INCU-Line® IL 750CR Premium 390-0966E
Additional standard stainless steel wire shelves, max. shelf load: 25 kg (reinforced stainless steel wire shelves, max. shelf load: 100 kg are also available)
Description Cat. No. Price Quantity
Additional shelf for IL 240CR 390-0967E
Additional shelf for IL 400CR 390-0968E
Additional shelf for IL 750CR 390-0969E