Would you like to log in again?
VWR® INCU-Line IL 23R cooled incubator
Incubator - 20%!
VWR® INCU-Line IL 23R cooled incubator
VALID UNTIL: 31/12/2017
For applications between 10 °C below ambient and + 50 °C
The IL 23R, with a chamber volume of 23 litres and Peltier technology for low energy consumption, offers an economical and space saving solution for microbiology applications at below room temperature. The transparent Plexiglas® door allows full visibility inside the incubator.
  • Digital PID controller with PT100 sensor and LED display
  • Excellent temperature accuracy of ±0,5 °C (at 37 °C)
  • Exterior WxDXH: 380x470x450 mm
  • Interior WxDxH: 294x300x270 mm
  • Supplied with two perforated shelves and temperature test certificate (data measured at 37 °C)
Temp. range (°C) 10 below ambient...50 °C
Temp. fluctuation (time) (°C) ±0,2 at 37 °C
Temp. (spatial) (°C) ±1,0 at 37 °C
Cap. (l) 23
Shelves supplied (max.) 2 (3)
Max. load per shelf/ chamber (kg) 3/9
Nominal power consumption (W) 80
Weight (kg) 19,2
Description Cat. No. Promo price Quantity
INCU-Line® IL 23R 390-0881E
Extra shelf for IL 23R 390-0483E