Would you like to log in again?
VWR® HiperSolv CHROMANORM® salts and acids for HPLC
VWR® HiperSolv CHROMANORM® salts and acids for HPLC
VALID UNTIL: 31/10/2017
HPLC salts and acids range to improve your HPLC separation.
In HPLC, for some applications, the mobile phase pH needs to be corrected to optimise separation. Buffer salts or acids are necessary, for instance, trifluoroacetic acid is important to improve the resolution in HPLC.
  • Very high quality
  • Available in small pack sizes
  • High UV transmission
HiPerSolv CHROMANORM®for HPLC Pk Cat. No. Promo price Quantity
Acetic acid ≥99,9% 250 ml 20108.230E
Ammonia 32% 100 ml 153312KE
Ammonium acetate ≥98% 250 g 153164RE
1-Heptanesulphonic acid sodium salt ≥99% 25 g 152782JE
1-Hexanesulphonic acid sodium salt ≥99% 25 g 152792LE
1-Octanesulphonic acid sodium salt ≥99% 25 g 152802TE
Orthophosphoric acid 85% 250 ml 153154DE
1-Pentanesulphonic acid sodium salt ≥99% 25 g 152812VE
Potassium dihydrogen phosphate ≥99,5% 500 g 153184UE
Sodium chloride 99,9% 500 g 153274VE
Sodium perchlorate monohydrate ≥99% 250 g 153233ME
Trifluoroacetic acid 100% 100 ml 153112EE