Would you like to log in again?
VWR® Easy-Read® environmentally friendly liquid-in-glass thermometers
VWR® Easy-Read® environmentally friendly liquid-in-glass thermometers
VALID UNTIL: 31/10/2017
Replace dangerous mercury thermometers with environmentally friendly Easy-Read®
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • EnviroKleen™ certified thermometers feature non toxic, non hazardous, biodegradable, Enviro-Safe® liquid
  • Accuracy without the potential hazards of mercury
  • Meets government, education and industry mandates for purchasing 'Green' products
Accuracy Range Divisions Length Immersion depth Pk Cat. No. Promo price Quantity
Partial immersion
±1 °C -10..+110 °C 1 °C 200 mm 50 mm 1 620-0783E
±1 °C, ±1,5 °C above 100 °C -10..+150 °C 1 °C 200 mm 50 mm 1 620-0781E
±1 °C, ±1,5 °C above 100 °C, ±2,5 °C above 200 °C, ±3 °C above 230 °C -10..+260 °C 1 °C 350 mm 76 mm 1 620-0784E
±1 °C -20..+110 °C 1 °C 300 mm 76 mm 1 620-0788E
±1 °C -20..+150 °C 1 °C 300 mm 76 mm 1 620-0786E
Total immersion
±0,5 °C -10..+110 °C 1 °C 200 mm Total 1 620-0782E
±1 °C, ±1,5 °C above ±100 °C, ±2 °C above 200 °C -10..+225 °C 1 °C 350 mm Total 1 620-0785E
±1 °C -20..+110 °C 1 °C 300 mm Total 1 620-0789E
±1 °C -20..+150 °C 1 °C 300 mm Total 1 620-0787E