Would you like to log in again?
Disposable PPE garments for use in your cleanroom - sterile and non sterile
Available as individual coveralls or kits (sterile)
Disposable PPE garments for use in your cleanroom  - sterile and non sterile
Protecting the operator as well as product and process in your cleanroom.
  • Coveralls (microporous) with hood Cat III type 4/5/6 (IEST -RP-CC003,3 - Level II)
  • Coveralls (SMS) with hood Cat III type 5/6, sterile and non sterile (IEST -RP-CC003,3 - Level I)
  • Also available as a sterile kit including coverall, hood, boots and sleeves
  • Each piece individually double packed
Size Pk Cat. No. Price Quantity
Coveralls with hood (non sterile), type 4/5/6
Small 50 113-8231E
Medium 50 113-8232E
Large 50 113-8233E
X-Large 50 113-8234E
XX-Large 50 113-8235E
XXX-Large 50 113-8236E
Cleanroom coveralls with hood (non sterile), type 5/6
Small 50 113-8225E
Medium 50 113-8226E
Large 50 113-8227E
X-Large 50 113-8228E
XX-Large 50 113-8229E
XXX-Large 50 113-8230E
Coveralls with hood (sterile)
Small 20 113-7881E
Medium 20 113-7882E
Large 20 113-7883E
X-Large 20 113-7884E
XX-Large 20 113-7885E
XXX-Large 20 113-7886E